4-(Bromomethyl)-3-fluorobenzonitrile 97%
Supplier: Ambeed
|
Danger
|
A247366-1G
A247366-25G
A247366-5G
A247366-10G
103811-762EA
18
USD
103811-762
103851-596
77090-926
77090-928
77090-930
77153-206
4-(Bromomethyl)-3-fluorobenzonitrile 97%
4-(Bromomethyl)-3-fluorobenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅BrFN MW: 214.04 g/mol Boiling Pt: 276.4 °C (760 mm Hg) Melting Pt: 77 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368341 CAS Number: 105942-09-4 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...