4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide 97%

Supplier: Ambeed
Danger

A229974-25G A229974-250MG A229974-5G
103819-512EA 62 USD
103819-512 77568-224 77892-380 77892-381
4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide 97%
4-(Bromomethyl)-1-methyl-1H-pyrazole hydrobromide

Precaución: For research use only. Inert atmosphere, store in freezer, under –20 °C.

Formula: C₅H₈Br₂N₂
Storage Temperature: Freezer
MDL Number: MFCD13195912
CAS Number: 528878-44-6
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR