4-Phenylbenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

A111554-100G A111554-10G A111554-25G A111554-500G A111554-5G A111554-1G
103797-858EA 11 USD
103797-858 103823-676 77284-126 77284-128 77284-130 77284-132 77284-134 77310-484
4-Phenylbenzyl bromide 98%
4-Phenylbenzyl bromide

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Boiling Pt: 333.4 °C (760 mm Hg)
Melting Pt: 83…86 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017869
CAS Number: 2567-29-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR