4-(Benzyloxy)cyclohexanone 96%

Supplier: Ambeed
Warning

A438192-100G A438192-10G A438192-250MG A438192-500G A438192-1G A438192-25G A438192-5G
103827-632EA 12 USD
103827-632 103838-696 76829-606 76829-608 76829-610 76829-612 76842-434 76842-436 76842-438
4-(Benzyloxy)cyclohexanone 96%
4-(Benzyloxy)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₂
MW: 204.2700 g/mol
Boiling Pt: 319.9 °C (760 mm Hg)
Flash Pt: 139.072 °C
Storage Temperature: Ambient
MDL Number: MFCD10697867
CAS Number: 2987-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR