4-(Benzyloxy)benzoic acid 98%

Supplier: Ambeed
Warning

A205314-100G A205314-10G A205314-25G A205314-5G A205314-1G
103817-624EA 6 USD
103817-624 103848-292 76785-474 76785-476 77086-502 77086-504 77302-954
4-(Benzyloxy)benzoic acid 98%
4-(Benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
Melting Pt: 189…192 °C
Flash Pt: 152.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00016527
CAS Number: 1486-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR