4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzyloxy)-5-carboxy-4-nitroansole, 5-(Benzyloxy)-2-carboxy-4-methoxynitrobenzene

A202613-1G A202613-25G A202613-5G A202613-10G A202613-250MG A202613-100G
103817-406EA 6 USD
103817-406 103848-050 77086-162 77086-164 77086-166 77164-638 77164-640 77683-564
4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid 98%
4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid

Precaución: For research use only.

Formula: C₁₅H₁₃NO₆
MW: 303.267 g/mol
Boiling Pt: 504.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114356
CAS Number: 60547-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR