4-(Benzyloxy)-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Succinic acid monobenzyl ester

A174844-25G A174844-5G A174844-100G A174844-10G A174844-250MG A174844-1G
103806-980EA 7 USD
103806-980 103845-280 77082-586 77082-588 77111-162 77111-164 77111-166 77301-354
4-(Benzyloxy)-4-oxobutanoic acid 97%
4-(Benzyloxy)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
Boiling Pt: 368.5 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 144.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00447717
CAS Number: 103-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR