4-(Benzyloxy)-3-hydroxybenzoic acid 97%

Supplier: Ambeed
Warning

A330975-250MG A330975-1G
103825-568EA 57 USD
103825-568 76828-332 77306-412
4-(Benzyloxy)-3-hydroxybenzoic acid 97%
4-(Benzyloxy)-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD20921505
CAS Number: 38853-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR