4-(Benzyloxy)-2-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A105026-250MG
A105026-25G
A105026-1G
A105026-5G
103796-818EA
23
USD
103796-818
103822-546
76780-776
76780-778
77309-410
77309-412
4-(Benzyloxy)-2-hydroxybenzaldehyde 97%
4-(Benzyloxy)-2-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Boiling Pt: 390.9 °C (760 mm Hg) Melting Pt: 78…80 °C Flash Pt: 234.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01075698 CAS Number: 52085-14-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...