4-(Aminomethyl)pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A385538-100MG
A385538-250MG
103826-784EA
87
USD
103826-784
103837-824
77572-146
77686-072
4-(Aminomethyl)pyrimidine 97%
4-(Aminomethyl)pyrimidine
Precaución: Keep in a dark place, inert atmosphere, 2…8 °C
Formula:
C₅H₇N₃ MW: 109.1292 g/mol Boiling Pt: 224.3 °C (760 mm Hg) Flash Pt: 112.07 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06796202 CAS Number: 45588-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...