4-(Aminomethyl)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A123143-1000G
A123143-100G
A123143-10G
A123143-25G
A123143-500G
76971-662EA
148.53
USD
76971-662
76971-664
76971-666
76971-668
76971-670
4-(Aminomethyl)benzoic acid 98%
4-(Aminomethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 323.1 °C (760 mm Hg) Melting Pt: ≥300 °C Flash Pt: 149.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010203 CAS Number: 56-91-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...