3,4-Dihydroxybenzylamine hydrobromide 97%

Supplier: Ambeed
Warning

A399015-1G A399015-5G A399015-250MG
103827-024EA 79 USD
103827-024 103838-062 76841-496 76841-498 77195-914
3,4-Dihydroxybenzylamine hydrobromide 97%
3,4-Dihydroxybenzylamine hydrobromide

Precaución: for Research Use Only

Formula: C₇H₁₀BrNO₂
Boiling Pt: 333.4 °C (760 mm Hg)
Melting Pt: 184…186 °C
Flash Pt: 155.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00012859
CAS Number: 16290-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR