4-(Aminomethyl)benzamide hydrochloride 98%

Supplier: Ambeed
Warning

A200114-100MG A200114-10G A200114-250MG A200114-1G A200114-25G A200114-5G
103817-164EA 44 USD
103817-164 103847-792 77164-538 77164-540 77164-542 77302-648 77302-650 77302-652
4-(Aminomethyl)benzamide hydrochloride 98%
4-(Aminomethyl)benzamide hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O
MW: 186.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07289058
CAS Number: 20188-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR