4-(Aminomethyl)aniline dihydrochloride 95%

Supplier: Ambeed
Warning

A151471-100G A151471-10G A151471-1G A151471-25G A151471-5G
103804-258EA 8 USD
103804-258 103842-410 77286-118 77286-120 77299-876 77299-878 77299-880
4-(Aminomethyl)aniline dihydrochloride 95%
4-(Aminomethyl)aniline dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₂Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD03160786
CAS Number: 54799-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR