4-(Aminomethyl)-7-methoxy-3,4-dihydroquinolin-2(1H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A138001-100MG A138001-250MG A138001-1G
103802-546EA 609 USD
103802-546 77010-212 77010-214 77298-948
4-(Aminomethyl)-7-methoxy-3,4-dihydroquinolin-2(1H)-one hydrochloride 97%
4-(Aminomethyl)-7-methoxy-3,4-dihydroquinolin-2(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClN₂O₂
MW: 242.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27920873
CAS Number: 1588440-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR