Pyridoxamine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride

A141540-100G A141540-1G A141540-5G
103803-006EA 7 USD
103803-006 76719-392 77316-172 77316-174
Pyridoxamine dihydrochloride 97%
Pyridoxamine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂·2HCl
MW: 241.12 g/mol
Boiling Pt: 460.1 °C (760 mm Hg)
Melting Pt: 224…226 °C
Flash Pt: 232.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00012808
CAS Number: 524-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR