4-Hydroxy-3-methoxybenzylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride

A207688-25G A207688-100G A207688-10G A207688-500G A207688-5G A207688-1000G
103848-496EA 5 USD
103848-496 77086-782 77287-842 77287-844 77287-846 77303-042 77748-302
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Boiling Pt: 292.9 °C (760 mm Hg)
Melting Pt: 219…221 °C
Flash Pt: 130.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00012864
CAS Number: 7149-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR