4-(Aminoethyl)-1-N-boc-piperidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminoethyl)-1-N-Boc-piperidine, 4-(2-Aminoethyl)piperidine, N1-BOC protected, tert-butyl 4-(2-aminoethyl)piperidine-1-carboxylate

A186462-1G A186462-25G A186462-5G A186462-250MG
103815-940EA 48 USD
103815-940 103846-522 77084-184 77084-186 77084-188 77685-437
4-(Aminoethyl)-1-N-boc-piperidine 95%
4-(Aminoethyl)-1-N-boc-piperidine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
Boiling Pt: 316 °C (760 mm Hg)
Flash Pt: 144.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038459
CAS Number: 146093-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR