4-(6-Bromo-3-pyridyl)morpholine 98%

Supplier: Ambeed
Warning

A210905-100MG A210905-5G A210905-250MG
103818-052EA 107 USD
103818-052 103848-744 77030-494 77030-496 77872-073
4-(6-Bromo-3-pyridyl)morpholine 98%
4-(6-Bromo-3-pyridyl)morpholine

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11869572
CAS Number: 952582-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR