4-(5-Methyl-3-phenylisoxazol-4-yl)benzene-1-sulfonyl chloride 97%

Supplier: Ambeed
Danger

A101187-10G A101187-250MG A101187-1G A101187-5G
103795-848EA 37 USD
103795-848 103821-532 77160-194 77160-196 77282-670 77282-672
4-(5-Methyl-3-phenylisoxazol-4-yl)benzene-1-sulfonyl chloride 97%
4-(5-Methyl-3-phenylisoxazol-4-yl)benzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₁₆H₁₂ClNO₃S
MW: 333.79 g/mol
Boiling Pt: 440.6±45.0 °C (760 mm Hg)
Flash Pt: 220.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26385552
CAS Number: 509074-26-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR