2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-yl)morpholine, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol eser, 2-(4-Morpholino)pyrimidine-5-boronic acid, pinacol ester
A208155-250MG
A208155-25G
A208155-5G
103817-858EA
49
USD
103817-858
103848-544
77030-406
77303-062
77303-064
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester 97%
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₂₂BN₃O₃ MW: 291.1538 g/mol Boiling Pt: 453.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD07368246 CAS Number: 957198-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...