4-(4-Piperidyl)-1-butanol ≥98%

Supplier: Ambeed
Warning

A182018-100MG A182018-250MG A182018-10G
103807-678EA 143 USD
103807-678 103846-048 77611-770 77611-772 77671-570
4-(4-Piperidyl)-1-butanol ≥98%
4-(4-Piperidyl)-1-butanol

Precaución: Keep in a dark place, inert atmosphere, room temperature

Formula: C₉H₁₉NO
MW: 157.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02178381
CAS Number: 57614-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR