1-(4-Aminophenyl)-4-methylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methylpiperazino)aniline, 4-(4-Methylpiperazin-1-yl)phenylamine, 4-(4-Methyl-1-piperazinyl)aniline

A100062-100G A100062-10G A100062-25G A100062-500G A100062-1G A100062-5G
103795-228EA 7 USD
103795-228 103820-950 76994-426 76994-428 76994-430 76994-432 77282-420 77282-422
1-(4-Aminophenyl)-4-methylpiperazine 97%
1-(4-Aminophenyl)-4-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
Boiling Pt: 351.5 °C (760 mm Hg)
Melting Pt: 89…91 °C
Flash Pt: 164 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00172703
CAS Number: 16153-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR