4-(4-Methylpiperazin-1-yl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methyl-1-piperazinyl)phenylboronic Acid

A842683-25G A842683-5G A842683-10G A842683-250MG A842683-1G
103841-210EA 108 USD
103841-210 76845-452 76845-454 77006-782 77006-784 77308-438
4-(4-Methylpiperazin-1-yl)phenylboronic acid 95%
4-(4-Methylpiperazin-1-yl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₂
MW: 220.08 g/mol
Boiling Pt: 406.2 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD11223486
CAS Number: 229009-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR