4-(4-Methoxyphenyl)butyric acid 97%

Supplier: Ambeed
Warning

A132991-100G A132991-10G A132991-1G A132991-25G A132991-5G
103801-810EA 5 USD
103801-810 77272-090 77272-092 77298-596 77314-400 77314-402
4-(4-Methoxyphenyl)butyric acid 97%
4-(4-Methoxyphenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 334.8 °C (760 mm Hg)
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00004404
CAS Number: 4521-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR