4-(4-Methoxyphenoxy)aniline 97%

Supplier: Ambeed
Warning

A133851-100G A133851-10G A133851-1G A133851-25G
76719-044EA 320.4 USD
76719-044 76719-046 76719-048 77314-618
4-(4-Methoxyphenoxy)aniline 97%
4-(4-Methoxyphenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.2478 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044105
CAS Number: 31465-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR