4-(4-Methoxynaphthalen-1-yl)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Genabilic Acid, Menbutone

A159798-100MG A159798-10G A159798-25G A159798-5G
103843-516EA 18 USD
103843-516 77273-266 77273-268 77300-420 77300-422
4-(4-Methoxynaphthalen-1-yl)-4-oxobutanoic acid 98%
4-(4-Methoxynaphthalen-1-yl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₄
MW: 258.26 g/mol
Boiling Pt: 511.5 °C (760 mm Hg)
Melting Pt: 170…174 °C
Flash Pt: 196.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00021539
CAS Number: 3562-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR