4-(4-Iodophenyl)butanoic acid 96%

Supplier: Ambeed
Warning

A130234-10G A130234-250MG A130234-1G A130234-5G A130234-25G
103801-404EA 21 USD
103801-404 76688-596 76688-598 77298-404 77298-406 77313-838
4-(4-Iodophenyl)butanoic acid 96%
4-(4-Iodophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₂
Boiling Pt: 351 °C (760 mm Hg)
Flash Pt: 166.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063353
CAS Number: 27913-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR