4-(4-Hydroxyphenyl)butanoic acid 98%

Supplier: Ambeed
Warning

A219756-10G A219756-250MG A219756-1G
103818-812EA 15 USD
103818-812 103849-692 76814-846 76814-848 77088-252
4-(4-Hydroxyphenyl)butanoic acid 98%
4-(4-Hydroxyphenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Boiling Pt: 364.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00033287
CAS Number: 7021-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR