4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde 97% (mixture of isomers)
Supplier: Ambeed
|
Warning
|
A150140-250MG
A150140-1G
A150140-5G
A150140-25G
A150140-100G
103804-134EA
29
USD
103804-134
103842-268
76798-614
77317-774
77317-776
77600-124
77879-040
4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde 97% (mixture of isomers)
4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₂₂O₂ Boiling Pt: 318.7 °C (760 mm Hg) Flash Pt: 93 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00019423 CAS Number: 31906-04-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...