4-(4-Ethylpiperazin-1-yl)phenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Ethylpiperazin-1-yl)aniline

A169399-100G A169399-10G A169399-1G A169399-25G A169399-5G
103806-382EA 13 USD
103806-382 103844-638 77011-548 77011-550 77081-710 77081-712 77081-714
4-(4-Ethylpiperazin-1-yl)phenylamine 98%
4-(4-Ethylpiperazin-1-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃
MW: 205.3000 g/mol
Boiling Pt: 364.4 °C (760 mm Hg)
Melting Pt: 76…78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00702246
CAS Number: 115619-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR