4-(4-Chlorophenyl)piperidin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Chlorophenyl)-4-hydroxypiperidine

A160694-10G A160694-250MG A160694-1G A160694-25G A160694-5G A160694-100G
103805-404EA 13 USD
103805-404 103843-640 76799-012 76799-014 77300-480 77300-482 77300-484 77667-256
4-(4-Chlorophenyl)piperidin-4-ol 98%
4-(4-Chlorophenyl)piperidin-4-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClNO
Boiling Pt: 344.5 °C (760 mm Hg)
Melting Pt: 139…141 °C
Flash Pt: 162.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00006001
CAS Number: 39512-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR