4-(4-Chlorophenyl)butyric acid 97%

Supplier: Ambeed
Warning

A110582-100G A110582-10G A110582-250MG A110582-25G A110582-1G A110582-5G
103797-686EA 19 USD
103797-686 103823-492 77284-012 77284-014 77284-016 77284-018 77310-292 77310-294
4-(4-Chlorophenyl)butyric acid 97%
4-(4-Chlorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.65 g/mol
Boiling Pt: 323 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06810494
CAS Number: 4619-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR