4-(4-Chlorophenyl)-2,6-diphenylpyrimidine 97%

Supplier: Ambeed
Warning

A268862-1G A268862-5G
103812-936EA 10 USD
103812-936 103852-840 77305-018 77305-020
4-(4-Chlorophenyl)-2,6-diphenylpyrimidine 97%
4-(4-Chlorophenyl)-2,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅ClN₂
Boiling Pt: 441.4±27.0 °C (760 mm Hg)
Melting Pt: 160…161 °C
Flash Pt: 253.3 °C
Storage Temperature: Ambient
MDL Number: MFCD29477692
CAS Number: 919301-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR