4-(4-Chlorobenzyl)-2H-phthalazin-1-one 95%

Supplier: Ambeed
Warning

A169097-100MG A169097-250MG A169097-1G A169097-25G A169097-5G
103806-354EA 147 USD
103806-354 103844-610 77286-738 77286-740 77300-962 77300-964 77300-966
4-(4-Chlorobenzyl)-2H-phthalazin-1-one 95%
4-(4-Chlorobenzyl)-2H-phthalazin-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClN₂O
Boiling Pt: 522.3 °C (760 mm Hg)
Flash Pt: 269.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02168779
CAS Number: 53242-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR