4-(4-Bromophenyl)pyridine 98%
Supplier: Ambeed
|
Warning
|
A147557-100MG
A147557-10G
A147557-250MG
A147557-1G
A147557-25G
A147557-5G
103803-798EA
20
USD
103803-798
103841-904
76812-054
76812-056
76812-058
77317-280
77317-282
77317-284
4-(4-Bromophenyl)pyridine 98%
4-(4-Bromophenyl)pyridine
Precaución: for Research Use Only
Formula:
C₁₁H₈BrN Boiling Pt: 318.7 °C (760 mm Hg) Flash Pt: 146.594 °C Storage Temperature: Ambient |
MDL Number:
MFCD00956748 CAS Number: 39795-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...