4-(4-Bromophenyl)piperidine 98%

Supplier: Ambeed
Warning

A213919-100MG A213919-250MG A213919-1G A213919-5G A213919-10G
103818-350EA 16 USD
103818-350 103849-060 76800-712 76800-714 77087-562 77303-310 77600-724
4-(4-Bromophenyl)piperidine 98%
4-(4-Bromophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN
MW: 240.15 g/mol
Boiling Pt: 307.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03426370
CAS Number: 80980-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR