4-(4-Bromophenyl)butyric acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Bromophenyl)butanoic acid

A129519-100G A129519-10G A129519-1G A129519-5G
103801-296EA 6 USD
103801-296 77285-254 77285-256 77298-352 77313-678
4-(4-Bromophenyl)butyric acid 97%
4-(4-Bromophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Boiling Pt: 351.1±17.0 °C (760 mm Hg)
Melting Pt: 71…72 °C
Flash Pt: 166.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00463170
CAS Number: 35656-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR