4-(4-Bromophenyl)-1-methyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A208001-1G
A208001-100MG
103817-846EA
121
USD
103817-846
103848-532
77086-832
77237-310
4-(4-Bromophenyl)-1-methyl-1H-pyrazole 97%
4-(4-Bromophenyl)-1-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₁₀H₉BrN₂ Boiling Pt: 345.3±25.0 °C (760 mm Hg) Flash Pt: 162.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD19322918 CAS Number: 1191616-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...