4-(4-Aminophenyl)morpholin-3-one 97%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Oxomorpholino)aniline

A280928-25G A280928-5G A280928-100G A280928-500G
103853-476EA 6 USD
103853-476 76837-168 76837-170 77114-054 77114-056
4-(4-Aminophenyl)morpholin-3-one 97%
4-(4-Aminophenyl)morpholin-3-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MW: 192.2145 g/mol
Flash Pt: 257.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08236742
CAS Number: 438056-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR