4-(4-Aminophenoxy)-N-methylpicolinamide 97%

Supplier: Ambeed
Warning

A269325-100G A269325-10G A269325-1G A269325-25G A269325-5G
103852-868EA 6 USD
103852-868 76787-160 76787-162 76787-164 77092-632 77092-634
4-(4-Aminophenoxy)-N-methylpicolinamide 97%
4-(4-Aminophenoxy)-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₃O₂
Boiling Pt: 476.8 °C (760 mm Hg)
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD08692047
CAS Number: 284462-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR