4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 95% (contains < 17% H₂O)
Supplier: Ambeed
|
Danger
|
Synonyms:
4-(4,6-Dimethoxy[1.3.5]triazin-2-yl)-4-methylmorpholinium chloride, DMTMM, 4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholin-4-ium chloride
A206238-25G
A206238-10G
103848-366EA
6
USD
103848-366
77086-612
77567-980
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride 95% (contains < 17% H₂O)
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride
Precaución: For research use only.
Formula:
C₁₀H₁₇ClN₄O₃ MW: 276.72 g/mol Melting Pt: 110…114 °C Flash Pt: 92.5 °C Storage Temperature: Freezer |
MDL Number:
MFCD03613550 CAS Number: 3945-69-5 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...