4-Pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Pyridine-4-boronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A175242-100G A175242-10G A175242-1G A175242-25G A175242-5G
103807-012EA 14 USD
103807-012 103845-316 76799-510 76799-512 77082-646 77082-648 77082-650
4-Pyridineboronic acid pinacol ester 97%
4-Pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Boiling Pt: 300.9 °C (760 mm Hg)
Melting Pt: 149…153 °C
Flash Pt: 135.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01319051
CAS Number: 181219-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR