2-Cyano-4-pyridineboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile
A124177-100MG
A124177-250MG
A124177-1G
A124177-5G
103800-494EA
169
USD
103800-494
77258-180
77258-182
77312-622
77312-624
2-Cyano-4-pyridineboronic acid pinacol ester 97%
2-Cyano-4-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BN₂O₂ MW: 230.07 g/mol Boiling Pt: 365.6±27.0 °C (760 mm Hg) Melting Pt: 60…61 °C Flash Pt: 174.9 °C Storage Temperature: Freezer |
MDL Number:
MFCD09607734 CAS Number: 741709-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...