Isoxazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 4-Isoxazoleboronic acid pinacol ester

A281292-100MG A281292-10G A281292-250MG A281292-1G A281292-5G
103813-544EA 17 USD
103813-544 103853-494 76787-402 76787-404 76787-406 76837-190 76837-192
Isoxazole-4-boronic acid pinacol ester 98%
Isoxazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₃
MW: 195.02 g/mol
Melting Pt: 110…115 °C
Flash Pt: 158.9 °C
Storage Temperature: Freezer
MDL Number: MFCD06657891
CAS Number: 928664-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR