4-Cyanophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyanobenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyanophenylboronic acid, pinacol ester

A268101-100G A268101-10G A268101-250MG A268101-1G A268101-5G A268101-25G
103812-894EA 6 USD
103812-894 103852-796 76722-976 76722-978 76722-980 77092-548 77092-550 77304-996
4-Cyanophenylboronic acid pinacol ester 97%
4-Cyanophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Boiling Pt: 345.8 °C (760 mm Hg)
Melting Pt: 95…99 °C
Flash Pt: 162.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03093897
CAS Number: 171364-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR