4-Carboxyphenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
A132897-100G
A132897-10G
A132897-500G
A132897-1G
A132897-25G
A132897-5G
103801-800EA
8
USD
103801-800
76719-014
76719-016
76719-018
77314-382
77314-384
77314-386
4-Carboxyphenylboronic acid pinacol ester 98%
4-Carboxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BO₄ MW: 248.08 g/mol Boiling Pt: 379.7 °C (760 mm Hg) Melting Pt: 228…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863710 CAS Number: 180516-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...