2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol ≥95%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzeneethanol

A406020-100MG A406020-250MG A406020-5G
103827-130EA 58 USD
103827-130 77594-230 77594-232 77672-982
2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol ≥95%
2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-ol

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Boiling Pt: 363.2±25.0 °C (760 mm Hg)
Flash Pt: 173.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18733921
CAS Number: 651030-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR