4-Formylbenzeneboronic acid pinacol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
A188411-100G
A188411-10G
A188411-250MG
A188411-500G
A188411-1G
A188411-25G
A188411-5G
103816-096EA
6
USD
103816-096
103846-692
76721-080
76721-082
76721-084
76721-086
77084-424
77084-426
77084-428
4-Formylbenzeneboronic acid pinacol ester 96%
4-Formylbenzeneboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BO₃ Boiling Pt: 341.3 °C (760 mm Hg) Melting Pt: 55…58 °C Flash Pt: 160.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972375 CAS Number: 128376-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...