4-Aminophenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester
A197547-100G
A197547-10G
A197547-500G
A197547-25G
A197547-5G
A197547-1G
103816-932EA
9
USD
103816-932
103847-552
76721-348
76721-350
76721-352
77085-562
77085-564
77302-552
4-Aminophenylboronic acid pinacol ester 98%
4-Aminophenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₈BNO₂ MW: 219.09 g/mol Boiling Pt: 340 °C (760 mm Hg) Melting Pt: 165…169 °C Flash Pt: 159.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093721 CAS Number: 214360-73-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...